Assay Details
Gene Symbol: Nipal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106995.1 | NP_001100465.1 | 4 - 5 | 510 | 63 | |
GenBank mRNA | FQ211728.1 | - | 4 - 5 | 579 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
