Assay Details
| Gene Symbol: Narf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039207.2 | NP_001034296.1 | 4 - 5 | 480 | 69 | |
| XM_006247899.2 | XP_006247961.1 | 4 - 5 | 513 | 69 | ||
| GenBank mRNA | BC110050.1 | - | 4 - 5 | 480 | 7454395 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Narf
Genomic Map