Assay Details
| Gene Symbol: Prss33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109027.1 | NP_001102497.1 | 2 - 3 | 90 | 78 | |
| XM_006245874.2 | XP_006245936.1 | 3 - 4 | 380 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prss33
Genomic Map