Assay Details
| Gene Symbol: B3gntl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015035.1 | NP_001015035.1 | 7 - 8 | 690 | 66 | |
| XM_006247904.2 | XP_006247966.1 | 7 - 8 | 740 | 66 | ||
| XM_008768503.1 | XP_008766725.1 | 7 - 8 | 740 | 66 | ||
| XM_008768504.1 | XP_008766726.1 | 7 - 8 | 740 | 66 | ||
| XM_008768505.1 | XP_008766727.1 | 7 - 8 | 740 | 66 | ||
| XM_008768506.1 | XP_008766728.1 | 7 - 8 | 740 | 66 | ||
| XM_008768507.1 | XP_008766729.1 | 7 - 8 | 740 | 66 | ||
| XM_008768508.1 | XP_008766730.1 | 7 - 8 | 740 | 66 | ||
| XM_008768509.1 | XP_008766731.1 | 7 - 8 | 740 | 66 | ||
| XM_008768510.1 | XP_008766732.1 | 5 - 6 | 443 | 66 | ||
| GenBank mRNA | AY634365.1 | - | 7 - 8 | 690 | 66 | |
| BC111711.1 | - | 7 - 8 | 664 | 7383845 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

B3gntl1
Genomic Map