Assay Details
| Gene Symbol: Adam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160228.1 | NP_001153700.1 | 9 - 10 | 999 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P096252 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P096257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10647 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P096257 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P096252 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P096257 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P096252 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P096252 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P096257 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P601971 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21753 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10732994 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732985 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732988 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732995 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732987 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732983 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732990 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732979 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732977 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732981 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732978 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732998 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732980 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732991 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732993 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732982 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732986 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732989 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732996 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10732984 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10732966 | - | - |
| Rat Genome 230 2.0 Array | 1395504_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395504_at | - | - |
| Rat Genome 230 2.0 Array | 1389151_at | - | - |
| Rat Genome 230 2.0 Array | 1385382_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385382_at | - | - |
| Rat Genome 230 2.0 Array | 1385665_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389151_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385665_at | - | - |
| Affymetrix HT Rat Focus Array | 1395504_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395504_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000003756.rn.1 | TC1000000445.rn.1 | RTA TC |
| Rat Expression Set 230 | 1385665_PM_at | - | - |
| Rat Expression Set 230 | 1389151_PM_at | - | - |
| Rat Expression Set 230 | 1385665_at | - | - |
| Rat Expression Set 230 | 1389151_at | - | - |
| Rat Expression Set 230 | 1385382_PM_at | - | - |
| Rat Expression Set 230 | 1395504_at | - | - |
| Rat Expression Set 230 | 1395504_PM_at | - | - |
| Rat Expression Set 230 | 1385382_at | - | - |
| Rat Genome U34 Set | rc_AA943114_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357629 | - | - |

Adam19
Genomic Map