Assay Details
Gene Symbol: Rpl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007599.2 | NP_001007600.1 | 3 - 4 | 250 | 94 | |
GenBank mRNA | BC058500.1 | - | 3 - 4 | 254 | 6921061 | 94 |
FQ221252.1 | - | 3 - 4 | 256 | 94 | ||
FQ221690.1 | - | 3 - 4 | 280 | 94 | ||
FQ222524.1 | - | 3 - 4 | 256 | 94 | ||
FQ222678.1 | - | 3 - 4 | 256 | 94 | ||
FQ223359.1 | - | 3 - 4 | 256 | 94 | ||
FQ224963.1 | - | 3 - 4 | 256 | 94 | ||
FQ228675.1 | - | 3 - 4 | 257 | 94 | ||
X58200.1 | - | 3 - 4 | 248 | 94 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
