Assay Details
| Gene Symbol: Rpl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007599.2 | NP_001007600.1 | 1 - 2 | 35 | 73 | |
| GenBank mRNA | BC058500.1 | - | 1 - 2 | 39 | 6921061 | 73 |
| FQ216928.1 | - | 1 - 2 | 41 | 73 | ||
| FQ221252.1 | - | 1 - 2 | 41 | 73 | ||
| FQ221690.1 | - | 1 - 2 | 65 | 73 | ||
| FQ221693.1 | - | 1 - 2 | 41 | 73 | ||
| FQ222524.1 | - | 1 - 2 | 41 | 73 | ||
| FQ222678.1 | - | 1 - 2 | 73 | |||
| FQ223359.1 | - | 1 - 2 | 41 | 73 | ||
| FQ224963.1 | - | 1 - 2 | 41 | 73 | ||
| X58200.1 | - | 1 - 2 | 33 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058500.1,NM_001007599.2,X58200.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl23
Genomic Map