Assay Details
| Gene Symbol: Sco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173374.1 | NP_001166845.1 | 2 - 3 | 355 | 98 | |
| GenBank mRNA | BC158882.1 | - | 2 - 3 | 290 | 98 | |
| CK359925.1 | - | 2 - 3 | 355 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sco1
Genomic Map