Assay Details
Gene Symbol: Maml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106997.1 | NP_001100467.1 | 2 - 3 | 2010 | 57 | |
XM_006246251.2 | XP_006246313.1 | 2 - 3 | 3284 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
