Assay Details
Gene Symbol: Osbpl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107044.1 | NP_001100514.1 | 6 - 7 | 741 | 59 | |
XM_006247299.2 | XP_006247361.1 | 7 - 8 | 798 | 59 | ||
XM_006247300.2 | XP_006247362.1 | 7 - 8 | 738 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
