Assay Details
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017045.1 | NP_058741.1 | 16 - 17 | 1847 | 80 | |
GenBank mRNA | D25233.1 | - | 16 - 17 | 1608 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
negative regulation of protein kinase activity
cell cycle arrest
Ras protein signal transduction
spermatogenesis
regulation of mitotic cell cycle
negative regulation of cell proliferation
chromatin modification
neuron differentiation
sister chromatid biorientation
neuron projection development
maintenance of mitotic sister chromatid cohesion
glial cell apoptotic process
skeletal muscle cell differentiation
neuron maturation
negative regulation of apoptotic process
enucleate erythrocyte differentiation
regulation of lipid kinase activity
myoblast differentiation
positive regulation of macrophage differentiation
negative regulation of cell cycle
positive regulation of mitotic metaphase/anaphase transition
negative regulation of smoothened signaling pathway
negative regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
digestive tract development
cell morphogenesis involved in neuron differentiation
negative regulation of epithelial cell proliferation
striated muscle cell differentiation
cell division
neuron apoptotic process
regulation of cell cycle
protein localization to chromosome, centromeric region
cellular response to xenobiotic stimulus
regulation of cohesin loading
negative regulation of transcription involved in G1/S transition of mitotic cell cycle
hepatocyte apoptotic process
negative regulation of G1/S transition of mitotic cell cycle
core promoter binding
RNA polymerase II activating transcription factor binding
DNA binding
protein binding
transcription factor binding
enzyme binding
kinase binding
ubiquitin protein ligase binding
identical protein binding
phosphoprotein binding
G1/S transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
negative regulation of protein kinase activity
cell cycle arrest
Ras protein signal transduction
spermatogenesis
regulation of mitotic cell cycle
negative regulation of cell proliferation
chromatin modification
neuron differentiation
sister chromatid biorientation
neuron projection development
maintenance of mitotic sister chromatid cohesion
glial cell apoptotic process
skeletal muscle cell differentiation
neuron maturation
negative regulation of apoptotic process
enucleate erythrocyte differentiation
regulation of lipid kinase activity
myoblast differentiation
positive regulation of macrophage differentiation
negative regulation of cell cycle
positive regulation of mitotic metaphase/anaphase transition
negative regulation of smoothened signaling pathway
negative regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
digestive tract development
cell morphogenesis involved in neuron differentiation
negative regulation of epithelial cell proliferation
striated muscle cell differentiation
cell division
neuron apoptotic process
regulation of cell cycle
protein localization to chromosome, centromeric region
cellular response to xenobiotic stimulus
regulation of cohesin loading
negative regulation of transcription involved in G1/S transition of mitotic cell cycle
hepatocyte apoptotic process
negative regulation of G1/S transition of mitotic cell cycle
core promoter binding
RNA polymerase II activating transcription factor binding
DNA binding
protein binding
transcription factor binding
enzyme binding
kinase binding
ubiquitin protein ligase binding
identical protein binding
phosphoprotein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P229079 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P229079 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P229079 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P229079 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P229079 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P328397 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700287464 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94980 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62564 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71721 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37071 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90966 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86473 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18449 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4274 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700287464 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701029985 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10785231 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785220 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785229 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785230 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785218 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785227 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785222 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785225 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785232 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785233 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785217 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785238 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785243 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785241 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785226 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785242 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785228 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785224 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785223 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785234 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785236 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785237 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785235 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785219 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785239 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785240 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10785216 | - | - |
Rat Genome 230 2.0 Array | 1388185_at | - | - |
Rat Genome 230 2.0 Array | 1388185_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388185_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388185_at | - | - |
Rat Expression Set 230 | 1388185_PM_at | - | - |
Rat Expression Set 230 | 1388185_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10785216 | - | - |
Rat Genome U34 Set | D25233cds_at | - | - |
Rat Genome U34 Set | rc_AI178012_at | - | - |
Rat Genome U34 Set | D25233UTR#1_g_at | - | - |
Rat Genome U34 Set | D25233UTR#1_at | - | - |
Rat Genome U34 Set | rc_AI071681_f_at | - | - |
Rat Genome U34 Set | D25233UTR#1_g_at | - | - |
Rat Genome U34 Set | D25233cds_at | - | - |
Rat Genome U34 Set | D25233UTR#1_at | - | - |
Rat Genome U34 Set | D25233UTR#1_at | - | - |
Rat Genome U34 Set | D25233UTR#1_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21157661 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371917 | - | - |
