Assay Details
| Gene Symbol: Ndfip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108390.2 | NP_001101860.2 | 2 - 3 | 584 | 78 | |
| XM_006252429.1 | XP_006252491.1 | 3 - 4 | 529 | 78 | ||
| GenBank mRNA | CO398212.1 | - | 2 - 3 | 367 | 78 | |
| DY565248.1 | - | 1 - 2 | 146 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndfip2
Genomic Map