Assay Details
| Gene Symbol: Lcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012044.1 | NP_001012044.1 | 15 - 16 | 1788 | 124 | |
| XM_006252318.2 | XP_006252380.1 | 15 - 16 | 1963 | 124 | ||
| XM_006252319.2 | XP_006252381.1 | 17 - 18 | 2162 | 124 | ||
| XM_006252320.2 | XP_006252382.1 | 17 - 18 | 2210 | 124 | ||
| XM_006252322.2 | XP_006252384.1 | 15 - 16 | 3369 | 124 | ||
| GenBank mRNA | BC083855.1 | - | 15 - 16 | 1788 | 7125327 | 124 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
T cell activation involved in immune response
protein kinase A signaling
cell migration
organ regeneration
regulation of intracellular protein transport
wound healing, spreading of cells
actin filament bundle assembly
molecular_function
calcium ion binding
identical protein binding
actin filament binding
T cell activation involved in immune response
protein kinase A signaling
cell migration
organ regeneration
regulation of intracellular protein transport
wound healing, spreading of cells
actin filament bundle assembly
molecular_function
calcium ion binding
identical protein binding
actin filament binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P032928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136206 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P032928 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P032928 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P032928 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P22080 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22360 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97778 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46140 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19062 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11425 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10781509 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781505 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781504 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781500 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781502 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781501 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781499 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781498 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10781496 | - | - |
| Rat Genome 230 2.0 Array | 1389210_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389210_at | - | - |
| Affymetrix HT Rat Focus Array | 1389210_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389210_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500005786.rn.1 | TC1500000842.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500005787.rn.1 | TC1500000842.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389210_at | - | - |
| Rat Expression Set 230 | 1389210_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781496 | - | - |
| Rat Genome U34 Set | rc_AI012958_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22095781 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376133 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376765 | - | - |

Lcp1
Genomic Map