Assay Details
Gene Symbol: Ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133589.2 | NP_598273.2 | 2 - 3 | 266 | 74 | |
GenBank mRNA | BC078747.1 | - | 2 - 3 | 266 | 7131278 | 74 |
X77236.1 | - | 2 - 3 | 231 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
