Assay Details
Gene Symbol: Atf6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002809.3 | NP_001002809.2 | 5 - 6 | 480 | 78 | |
XM_006255983.1 | XP_006256045.1 | 5 - 6 | 629 | 78 | ||
GenBank mRNA | BC098631.1 | - | 5 - 6 | 471 | 7103893 | 78 |
FM061416.1 | - | 5 - 6 | 480 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
