Assay Details
| Gene Symbol: Srms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011961.1 | NP_001011961.1 | 4 - 5 | 926 | 72 | |
| GenBank mRNA | BC090006.1 | - | 4 - 5 | 926 | 7323399 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Srms
Genomic Map