Assay Details
| Gene Symbol: Pde4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276765.1 | NP_001263694.1 | 10 - 11 | 1071 | 66 | |
| XM_006252868.2 | XP_006252930.1 | 11 - 12 | 1313 | 66 | ||
| XM_006252869.2 | XP_006252931.1 | 9 - 10 | 1118 | 66 | ||
| GenBank mRNA | L27061.1 | - | 8 - 9 | 66 | ||
| M25347.1 | - | 2 - 3 | 210 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde4c
Genomic Map