Assay Details
Gene Symbol: Ddx26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773666.1 | XP_008771888.1 | 11 - 12 | 1810 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
