Assay Details
Gene Symbol: Ddx26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773666.1 | XP_008771888.1 | 15 - 16 | 2571 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
