Assay Details
| Gene Symbol: Aldoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177305.1 | NP_001170776.1 | 1 - 2 | 37 | 79 | |
| GenBank mRNA | CO398643.1 | - | 1 - 2 | 30 | 79 | |
| FQ216189.1 | - | 1 - 2 | 45 | 79 | ||
| FQ216820.1 | - | 1 - 2 | 41 | 79 | ||
| M14420.1 | - | 1 - 2 | 37 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the M14420.1, NM_001177305.1 transcripts.
Set Membership: |
|

Aldoa
Genomic Map