Assay Details
| Gene Symbol: Mastl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107369.1 | NP_001100839.1 | 1036 | 74 | ||
| XM_006254340.2 | XP_006254402.1 | 1098 | 74 | |||
| XM_006254341.2 | XP_006254403.1 | 960 | 74 | |||
| XM_006254342.2 | XP_006254404.1 | 1102 | 74 | |||
| GenBank mRNA | FQ224658.1 | - | 1195 | 74 | ||
More Information
Set Membership: |
|

Mastl
Genomic Map