Assay Details
| Gene Symbol: Zbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133564.1 | NP_598248.1 | 2 - 3 | 271 | 118 | |
| XM_006235674.2 | XP_006235736.1 | 3 - 4 | 4114 | 118 | ||
| GenBank mRNA | AJ302054.1 | - | 2 - 3 | 271 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zbp1
Genomic Map