Assay Details
Gene Symbol: Gtpbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100815.1 | NP_001094285.1 | 5 - 6 | 608 | 69 | |
GenBank mRNA | CV103632.1 | - | 3 - 4 | 274 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
