Assay Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023093.1 | NP_075581.1 | 8 - 9 | 1222 | 83 | |
| GenBank mRNA | BC061830.1 | - | 8 - 9 | 1222 | 5599635 | 83 |
| FQ214146.1 | - | 7 - 8 | 1064 | 83 | ||
| FQ225416.1 | - | 3 - 4 | 537 | 83 | ||
| FQ225735.1 | - | 3 - 4 | 537 | 83 | ||
| FQ227531.1 | - | 3 - 4 | 537 | 83 | ||
| FQ227720.1 | - | 2 - 3 | 357 | 83 | ||
| X56133.1 | - | 1 - 2 | 219 | 83 | ||
More Information
Set Membership: |
|

Atp5a1
Genomic Map