Assay Details
| Gene Symbol: Rbbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134417.1 | NP_001127889.1 | 9 - 10 | 1076 | 69 | |
| XM_006254407.1 | XP_006254469.1 | 9 - 10 | 1220 | 69 | ||
| XM_006254408.2 | XP_006254470.1 | 9 - 10 | 1242 | 69 | ||
| XM_008771938.1 | XP_008770160.1 | 9 - 10 | 1299 | 69 | ||
| XM_008771939.1 | XP_008770161.1 | 9 - 10 | 1145 | 69 | ||
| XM_008771940.1 | XP_008770162.1 | 9 - 10 | 1121 | 69 | ||
| XM_008771941.1 | XP_008770163.1 | 9 - 10 | 1197 | 69 | ||
| XM_008771942.1 | XP_008770164.1 | 9 - 10 | 1302 | 69 | ||
| GenBank mRNA | BC162012.1 | - | 9 - 10 | 1076 | 9091712 | 69 |
| FQ209715.1 | - | 9 - 10 | 1128 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbbp8
Genomic Map