Assay Details
| Gene Symbol: Nceh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127524.2 | NP_001120996.1 | 3 - 4 | 488 | 84 | |
| GenBank mRNA | BC166533.1 | - | 3 - 4 | 488 | 7190480 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Nceh1
Genomic Map