Assay Details
| Gene Symbol: Usp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008863.3 | NP_001008863.2 | 8 - 9 | 977 | 144 | |
| XM_008770560.1 | XP_008768782.1 | 11 - 12 | 2340 | 144 | ||
| GenBank mRNA | BI282345.1 | - | 3 - 4 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P033852 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P795568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P033852 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P795568 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P795568 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P033852 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P033852 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P795568 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51891 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22171 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73426 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22046 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63114 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55449 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7241 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10779086 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779095 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779097 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779082 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779094 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779084 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779088 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779090 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779085 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779098 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779083 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779096 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779087 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779089 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10779081 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10779093 | - | - |
| Rat Genome 230 2.0 Array | 1372356_at | - | - |
| Rat Genome 230 2.0 Array | 1372356_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372356_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372356_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500000404.rn.1 | TC1500000039.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372356_at | - | - |
| Rat Expression Set 230 | 1372356_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779081 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779093 | - | - |
| Rat Genome U34 Set | rc_AI102472_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22133006 | - | - |
| ABI Rat Genome Survey Microarray | 22346206 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040306 | - | - |

Usp54
Genomic Map