Assay Details
| Gene Symbol: Kcnk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039516.2 | NP_001034605.1 | 1 - 2 | 526 | 72 | |
| GenBank mRNA | AM229406.1 | - | 1 - 2 | 181 | 72 | |
| BC129068.1 | - | 1 - 2 | 526 | 7387039 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P776390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55455 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P776390 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P776390 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P776390 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50700 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22812 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82901 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6384 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10779188 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779190 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779185 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779186 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779187 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779189 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10779168 | - | - |
| Rat Genome 230 2.0 Array | 1371898_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371898_at | - | - |
| Affymetrix HT Rat Focus Array | 1371898_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371898_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500000650.rn.1 | TC1500000054.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500000642.rn.1 | TC1500000054.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500000648.rn.1 | TC1500000054.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500000643.rn.1 | TC1500000054.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500000649.rn.1 | TC1500000054.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371898_PM_at | - | - |
| Rat Expression Set 230 | 1371898_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779168 | - | - |
| Rat Genome U34 Set | rc_AA800928_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21592217 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365014 | - | - |

Kcnk5
Genomic Map