Assay Details
| Gene Symbol: Nsun2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108403.1 | NP_001101873.1 | 8 - 9 | 967 | 64 | |
| XM_006227792.2 | XP_006227854.1 | 7 - 8 | 902 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsun2
Genomic Map