Assay Details
| Gene Symbol: Ube2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251700.2 | XP_006251762.1 | 3 - 4 | 369 | 130 | |
| GenBank mRNA | FQ225411.1 | - | 3 - 4 | 284 | 130 | |
| Gene Symbol: Ube2e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC079134.1 | - | 486 | 130 | ||
| BC099169.1 | - | 460 | 130 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ube2e1
Genomic Map