Assay Details
| Gene Symbol: Nkiras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107252.2 | NP_001100722.1 | 3 - 4 | 396 | 87 | |
| XM_006251698.2 | XP_006251760.1 | 2 - 3 | 370 | 87 | ||
| XM_008770514.1 | XP_008768736.1 | 3 - 4 | 524 | 87 | ||
| GenBank mRNA | BC169080.1 | - | 3 - 4 | 396 | 5623298 | 87 |
| FQ211880.1 | - | 281 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nkiras1
Genomic Map