Assay Details
| Gene Symbol: Ptpn20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080149.1 | NP_001073618.1 | 4 - 5 | 414 | 97 | |
| XM_006252765.2 | XP_006252827.1 | 5 - 6 | 487 | 97 | ||
| XM_006252767.2 | XP_006252829.1 | 5 - 6 | 454 | 97 | ||
| XM_008771099.1 | XP_008769321.1 | 5 - 6 | 440 | 97 | ||
| XM_008771100.1 | XP_008769322.1 | 5 - 6 | 487 | 97 | ||
| GenBank mRNA | BC129117.1 | - | 4 - 5 | 414 | 7114504 | 97 |
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn20b
Genomic Map