Assay Details
| Gene Symbol: LOC685179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001055673.5 | XP_001055673.1 | 19 - 20 | 1933 | 66 | |
| XM_001055738.5 | XP_001055738.1 | 18 - 19 | 1840 | 66 | ||
| XM_001055795.5 | XP_001055795.1 | 18 - 19 | 1841 | 66 | ||
| XM_002729767.4 | XP_002729813.2 | 19 - 20 | 1934 | 66 | ||
| XM_006240805.2 | XP_006240867.1 | 19 - 20 | 1931 | 66 | ||
| XM_006240806.2 | XP_006240868.1 | 19 - 20 | 1913 | 66 | ||
| XM_006240807.2 | XP_006240869.1 | 18 - 19 | 1838 | 66 | ||
| XM_006240808.2 | XP_006240870.1 | 18 - 19 | 1820 | 66 | ||
| XM_008765050.1 | XP_008763272.1 | 19 - 20 | 1934 | 66 | ||
| XM_008765051.1 | XP_008763273.1 | 19 - 20 | 2778 | 66 | ||
More Information
Set Membership: |
|

LOC685179
Genomic Map