Assay Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134364.1 | NP_599191.1 | 5 - 6 | 814 | 106 | |
| GenBank mRNA | BC099743.1 | - | 5 - 6 | 797 | 7937989 | 106 |
| DQ403101.1 | - | 2 - 3 | 294 | 106 | ||
| FQ214851.1 | - | 2 - 3 | 306 | 106 | ||
| FQ216116.1 | - | 3 - 4 | 394 | 106 | ||
| FQ230301.1 | - | 3 - 4 | 433 | 106 | ||
| FQ231816.1 | - | 1 - 2 | 193 | 106 | ||
| M19044.1 | - | 4 - 5 | 630 | 106 | ||
| M57634.1 | - | 2 - 3 | 279 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5b
Genomic Map