Assay Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134364.1 | NP_599191.1 | 6 - 7 | 972 | 85 | |
| GenBank mRNA | BC099743.1 | - | 6 - 7 | 955 | 7937989 | 85 |
| DQ403101.1 | - | 3 - 4 | 452 | 85 | ||
| FQ214851.1 | - | 3 - 4 | 464 | 85 | ||
| FQ216116.1 | - | 4 - 5 | 552 | 85 | ||
| FQ230301.1 | - | 4 - 5 | 591 | 85 | ||
| FQ231816.1 | - | 2 - 3 | 351 | 85 | ||
| M19044.1 | - | 5 - 6 | 788 | 85 | ||
| M57634.1 | - | 3 - 4 | 437 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp5b
Genomic Map