Assay Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134364.1 | NP_599191.1 | 7 - 8 | 1096 | 57 | |
| GenBank mRNA | BC099743.1 | - | 7 - 8 | 1079 | 7937989 | 57 |
| DQ403101.1 | - | 4 - 5 | 576 | 57 | ||
| FQ214851.1 | - | 4 - 5 | 588 | 57 | ||
| FQ231816.1 | - | 3 - 4 | 475 | 57 | ||
| M19044.1 | - | 6 - 7 | 912 | 57 | ||
| M57634.1 | - | 4 - 5 | 561 | 57 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp5b
Genomic Map