Assay Details
| Gene Symbol: Fbxl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248906.2 | XP_006248968.1 | 1 - 2 | 87 | 68 | |
| XM_006248907.2 | XP_006248969.1 | 1 - 2 | 131 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006248906.2, XM_006248907.2 transcripts.
Set Membership: |
|

Fbxl18
Genomic Map