Assay Details
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047849.1 | NP_001041314.1 | 4 - 5 | 1460 | 66 | |
GenBank mRNA | BC082092.1 | - | 1 - 2 | 306 | 66 | |
BC161809.1 | - | 3 - 4 | 746 | 8374387 | 66 | |
DQ480750.1 | - | 4 - 5 | 1460 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
