Assay Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047849.1 | NP_001041314.1 | 5 - 6 | 1535 | 87 | |
| GenBank mRNA | BC082092.1 | - | 2 - 3 | 381 | 87 | |
| BC161809.1 | - | 4 - 5 | 821 | 8374387 | 87 | |
| DQ480750.1 | - | 5 - 6 | 1535 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat6
Genomic Map