Assay Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047849.1 | NP_001041314.1 | 6 - 7 | 1625 | 66 | |
| GenBank mRNA | BC082092.1 | - | 3 - 4 | 471 | 66 | |
| BC161809.1 | - | 5 - 6 | 911 | 8374387 | 66 | |
| DQ480750.1 | - | 6 - 7 | 1625 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat6
Genomic Map