Assay Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047849.1 | NP_001041314.1 | 8 - 9 | 1840 | 85 | |
| GenBank mRNA | BC082092.1 | - | 5 - 6 | 686 | 85 | |
| BC161809.1 | - | 7 - 8 | 1126 | 8374387 | 85 | |
| DQ480750.1 | - | 8 - 9 | 1840 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat6
Genomic Map