Assay Details
Gene Symbol: Mfhas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107316.1 | NP_001100786.1 | 1 - 2 | 2986 | 57 | |
XM_006253224.2 | XP_006253286.1 | 1 - 2 | 3069 | 57 | ||
XM_006253225.2 | XP_006253287.1 | 1 - 2 | 3069 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
