Assay Details
Gene Symbol: Mfhas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107316.1 | NP_001100786.1 | 2 - 3 | 3112 | 85 | |
XM_006253225.2 | XP_006253287.1 | 2 - 3 | 3195 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P283790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68715 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P283790 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P283790 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P283790 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P283790 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51419 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16527 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10792026 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792027 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10792024 | - | - |
Rat Genome 230 2.0 Array | 1398648_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398648_at | - | - |
Rat Genome 230 2.0 Array | 1384657_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393738_PM_s_at | - | - |
Rat Genome 230 2.0 Array | 1393738_s_at | - | - |
Rat Genome 230 2.0 Array | 1384657_at | - | - |
Affymetrix HT Rat Focus Array | 1393738_s_at | - | - |
Affymetrix HT Rat Focus Array | 1384657_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393738_PM_s_at | - | - |
Affymetrix HT Rat Focus Array | 1398648_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398648_at | - | - |
Affymetrix HT Rat Focus Array | 1384657_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600013058.rn.1 | TC1600001757.rn.1 | RTA TC |
Rat Expression Set 230 | 1384657_PM_at | - | - |
Rat Expression Set 230 | 1398648_at | - | - |
Rat Expression Set 230 | 1393738_s_at | - | - |
Rat Expression Set 230 | 1384657_at | - | - |
Rat Expression Set 230 | 1393738_PM_s_at | - | - |
Rat Expression Set 230 | 1398648_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792024 | - | - |
Rat Genome U34 Set | U47314_at | - | - |
Rat Genome U34 Set | rc_AI071532_at | - | - |
Rat Genome U34 Set | U47314_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357770 | - | - |
