Assay Details
| Gene Symbol: Supt3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244587.2 | XP_006244649.2 | 10 - 11 | 1744 | 74 | |
| XM_008766906.1 | XP_008765128.1 | 10 - 11 | 1744 | 74 | ||
| XM_008766907.1 | XP_008765129.1 | 10 - 11 | 1217 | 74 | ||
| XM_008766908.1 | XP_008765130.1 | 10 - 11 | 1113 | 74 | ||
| XM_008766909.1 | XP_008765131.1 | 10 - 11 | 1074 | 74 | ||
| XM_008766910.1 | XP_008765132.1 | 9 - 10 | 987 | 74 | ||
| XM_008766911.1 | XP_008765133.1 | 9 - 10 | 972 | 74 | ||
More Information
Set Membership: |
|

Supt3h
Genomic Map