Assay Details
| Gene Symbol: Serpinb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007732.1 | NP_001007733.1 | 5 - 6 | 743 | 133 | |
| XM_006253872.2 | XP_006253934.1 | 5 - 6 | 712 | 133 | ||
| GenBank mRNA | BC079063.1 | - | 5 - 6 | 743 | 7121006 | 133 |
| FQ226618.1 | - | 2 - 3 | 278 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb9
Genomic Map