Assay Details
| Gene Symbol: Ripk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107350.1 | NP_001100820.1 | 10 - 11 | 2051 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
apoptotic process
positive regulation of cell death
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
response to tumor necrosis factor
peptidyl-serine autophosphorylation
positive regulation of phosphorylation
positive regulation of apoptotic process
positive regulation of programmed cell death
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAPK cascade
cellular protein catabolic process
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
protein autophosphorylation
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of transferase activity
positive regulation of necroptotic process
T cell apoptotic process
necroptotic process
regulation of ATP:ADP antiporter activity
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
extrinsic apoptotic signaling pathway
ripoptosome assembly
necroptotic signaling pathway
ripoptosome assembly involved in necroptotic process
amyloid fibril formation
regulation of reactive oxygen species metabolic process
negative regulation of extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
death receptor binding
protein binding
ATP binding
ubiquitin protein ligase binding
protein complex binding
identical protein binding
death domain binding
positive regulation of protein phosphorylation
apoptotic process
positive regulation of cell death
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
response to tumor necrosis factor
peptidyl-serine autophosphorylation
positive regulation of phosphorylation
positive regulation of apoptotic process
positive regulation of programmed cell death
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAPK cascade
cellular protein catabolic process
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
protein autophosphorylation
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of transferase activity
positive regulation of necroptotic process
T cell apoptotic process
necroptotic process
regulation of ATP:ADP antiporter activity
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
extrinsic apoptotic signaling pathway
ripoptosome assembly
necroptotic signaling pathway
ripoptosome assembly involved in necroptotic process
amyloid fibril formation
regulation of reactive oxygen species metabolic process
negative regulation of extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
death receptor binding
protein binding
ATP binding
ubiquitin protein ligase binding
protein complex binding
identical protein binding
death domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P156830 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P722994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131931 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P722994 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P156830 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P722994 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P156830 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P156830 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P722994 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P156830 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13219 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100505 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37788 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48866 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10798113 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798118 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798111 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798114 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798116 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798109 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798115 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798117 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10798108 | - | - |
| Rat Genome 230 2.0 Array | 1371529_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391543_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391543_at | - | - |
| Rat Genome 230 2.0 Array | 1371529_at | - | - |
| Affymetrix HT Rat Focus Array | 1391543_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391543_at | - | - |
| Affymetrix HT Rat Focus Array | 1371529_at | - | - |
| Affymetrix HT Rat Focus Array | 1371529_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700011444.rn.1 | TC1700001651.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700011442.rn.1 | TC1700001651.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700011445.rn.1 | TC1700001651.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700011446.rn.1 | TC1700001651.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371529_PM_at | - | - |
| Rat Expression Set 230 | 1371529_at | - | - |
| Rat Expression Set 230 | 1391543_at | - | - |
| Rat Expression Set 230 | 1391543_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10798108 | - | - |
| Rat Genome U34 Set | rc_AI007920_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20966050 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350981 | - | - |

Ripk1
Genomic Map