Assay Details
Gene Symbol: Riok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100511.1 | NP_001093981.1 | 14 - 15 | 1560 | 63 | |
XM_006253789.2 | XP_006253851.1 | 14 - 15 | 1629 | 63 | ||
GenBank mRNA | BC079173.1 | - | 14 - 15 | 1560 | 63 | |
CO565410.1 | - | 2 - 3 | 176 | 63 | ||
FQ209749.1 | - | 14 - 15 | 1614 | 63 | ||
FQ234746.1 | - | 11 - 12 | 1027 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
