Assay Details
Gene Symbol: Mrps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191605.3 | NP_001178534.1 | 1 - 2 | 119 | 80 | |
GenBank mRNA | FQ224224.1 | - | 1 - 2 | 53 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191605.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
