Assay Details
| Gene Symbol: Ccr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227922.2 | XP_006227984.1 | 2 - 3 | 370 | 107 | |
| XM_008758755.1 | XP_008756977.1 | 1 - 2 | 137 | 107 | ||
| GenBank mRNA | FQ225692.1 | - | 1 - 2 | 137 | 107 | |
| FQ226017.1 | - | 1 - 2 | 137 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006227922.2, XM_008758755.1 transcripts.
Set Membership: |
|

Ccr6
Genomic Map