Assay Details
| Gene Symbol: Pabpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106208.2 | NP_001099678.1 | 1 | 2604 | 113 | |
| GenBank mRNA | BC168959.1 | - | 1 | 2604 | 7135469 | 113 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pabpc6
Genomic Map